missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SYTO™ Blue Fluorescent Nucleic Acid Stain Sampler Kit (SYTO™ dyes 40, 41, 42, 45)

Product Code. 10297192
Click to view available options
Quantity:
1 kit
Unit Size:
Each
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 10297192

Brand: Invitrogen™ S11350

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Includes

The kit contains 50μL of each SYTO dye.

Includes cell-permeant, blue-fluorescent SYTO dyes

The SYTO Blue Fluorescent Nucleic Acid Stain Sampler Kit contains a collection of cell-permeant, blue-fluorescent SYTO nucleic acid stains (SYTO dyes 40, 41, 42, 45). Because the dyes may demonstrate different staining behaviors with various tissues and cells, it may be necessary to test the dyes to find the optimal dye for a specific application. The kit contains 50 μL of each SYTO dye.

Any physiological buffer between pH 7.0–8.0, including PBS, can be used to dilute the SYTO dyes for the staining solution.

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Color Blue
Content And Storage The kit contains 50 μL of each SYTO dye.
  • Store in freezer -5°C to -30°C and protect from light.
  • Excitation Wavelength Range various
    Dye Type Cell-Permeant
    For Use With (Equipment) Fluorescence Microscope
    Quantity 1 kit
    Detection Method Fluorescence
    Product Line SYTO™
    Shipping Condition Room Temperature
    Label Type Fluorescent Dye
    Product Type Nucleic Acid Stain
    SubCellular Localization Nucleic Acids
    Show More Show Less
    compliance-icons
    Product Identifier
    • SYTO Blue Fluorescent Nucleic Acid Stain Sampler Kit (SYTO dyes 40, 41, 42, 45)
    Signal Word
    • Warning
    Hazard Category
    • Serious eye damage/eye irritation Category 2
    • Flammable liquid Category 3
    • Specific target organ toxicity Category 3
    Hazard Statement
    • H335-May cause respiratory irritation.
    Precautionary Statement
    • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
    • P264-Wash hands thoroughly after handling.
    • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
    • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    • P312-Call a POISON CENTER/doctor/if you feel unwell.
    • P332+P313-If skin irritation occurs: Get medical advice/attention.
    • P337+P313-If eye irritation persists: Get medical advice/attention.
    Supplemental information
    • MIXTURE LIST-Contains : Dimethyl sulfoxide,SYTO 45 dye, SYTO 44dye

    For Research Use Only. Not for use in diagnostic procedures.

    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.