Learn More
Invitrogen™ Rhod-2, AM, cell permeant
Cells may be loaded with AM ester forms of these calcium indicators by adding dissolved indicator directly to dishes containing cultured cells
£516.00 - £588.00
Specifications
Content And Storage | Store in freezer -5°C to -30°C and protect from light. |
---|---|
Detection Method | Fluorescent |
For Use With (Application) | Cell Viability and Proliferation |
For Use With (Equipment) | Fluorescence Microscope |
Product Type | Stain |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10217112
|
Invitrogen™
R1244 |
1mg |
£516.00
1mg |
Estimated Shipment: 29-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11533677
|
Invitrogen™
R1245MP |
20 x 50μg |
£588.00
1000µg |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Labeled calcium indicators are molecules that exhibit increase in fluorescence upon binding Ca2+. They have uses in many calcium signaling investigations, including measuring Ca2+ in cells and tissues that have high levels of autofluorescence and also for detecting Ca2+ release generated by photoreceptors and photoactivatable chelators.
- Fluorescence signal from these cells is generally measured using fluorescence microscopy
- Label (Ex/Em of Ca2+-bound form): Rhod-2 (552/581nm)
- Fluorescence intensity increase upon binding Ca2+: >100 fold
- Kd for Ca2+ in the absence of Mg2+, in buffer: approximately 570nM
- Exhibit fluorescence increase upon binding Ca2+ with little shift in wavelength
- In addition, BAPTA-based indicators such as these bind various heavy metal cations (e.g., Mn2+, Zn2+, Pb2+) with substantially higher affinity than Ca2+
- Perturbations to calcium measurements caused by presence of these ions can be controlled using the heavy metal-selective chelator TPEN
Calcium Detection, Cell Analysis, Cell Viability, Proliferation and Function, Ionic Homeostasis and Signaling
Order Info
Shipping Condition: Room temperature
Specifications
Store in freezer -5°C to -30°C and protect from light. | |
Cell Viability and Proliferation | |
Stain | |
Room Temperature |
Fluorescent | |
Fluorescence Microscope | |
Fluorescent Dye-Based |
For Research Use Only. Not for use in diagnostic procedures.