Learn More
Invitrogen™ Molecular Probes™ PRO-Q™ Diamond Phosphoprotein Gel Staining Kit
Can detect as little as 1-16ng of phosphoprotein per band with a linear signal over three orders of magnitude
Brand:  Invitrogen™ MPP33302
Code : 50
Additional Details : Weight : 1.00000kg
 View more versions of this product
Product Code. 10768414
  • £1524.39 / 1 set
Estimated Shipment
06-02-2018
Add to basket
For Research Use Only. All usage must comply with product instructions.
Disclaimers:
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Specifications
Protein Gel Electrophoresis, Protein Gel Staining and Imaging, Proteins, Expression, Isolation and Analysis
5L of the Pro-Q™ Diamond Phosphoprotein Gel Stain and 40μL of the PeppermintStick™ phosphoprotein molecular weight standards
Videos