missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Pro-Q™ Diamond Phosphoprotein Gel Stain
Pro-Q Diamond Phosphoprotein Gel Stain provides a convenient method for selectively staining phosphoproteins in acrylamide gels, without the need for blotting or use of phosphoprotein-specific antibodies.
£160.00 - £2140.00
Specifications
Content And Storage | Store at room temperature and protect from light. |
---|---|
Detection Location | In-Gel Detection |
Detection Method | Fluorescence |
Label or Dye | Pro-Q Diamond |
Product Type | Phosphoprotein Gel Stain |
Product Code | Brand | Description | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Price | Quantity & Availability | |||||
11549296
|
Invitrogen™
P33301 |
Pro-Q™ Diamond Phosphoprotein Gel Stain, 200 mL |
£160.00
200mL |
Estimated Shipment: 12-04-2024
Log in to see stock. |
|||||
10534433
|
Invitrogen™
P33300 |
Pro-Q™ Diamond Phosphoprotein Gel Stain, 1 L |
£495.00
1 L |
Estimated Shipment: 16-04-2024
Log in to see stock. |
|||||
11406347
|
Invitrogen™
P33302 |
Pro-Q™ Diamond Phosphoprotein Gel Stain, 5 L |
£2140.00
5 L |
Estimated Shipment: 16-04-2024
Log in to see stock. |
|||||
Description
Pro-Q Diamond Phosphoprotein Gel Stain provides a convenient method for selectively staining phosphoproteins in acrylamide gels, without the need for blotting or use of phosphoprotein-specific antibodies. It is ideal for the identification of kinase targets in signal transduction pathways and for phosphoproteomic studies.
Pro-Q Diamond Phosphoprotein Gel Stain provides a convenient method for selectively staining phosphoproteins in acrylamide gels, without the need for blotting or use of phosphoprotein specific antibodies. It is ideal for the identification of kinase targets in signal transduction pathways and for phosphoproteomic studies.Features include:
• Sensitive—detection level down to 1–16 ng of phosphoprotein per band
• Convenient—selectively stain phosphoproteins in acrylamide gels, without the need for blotting or phosphoprotein specific antibodies and western blot analysis
• Linear signal—exhibits excellent linearity over three orders of magnitude, ideal for quantitative western blotting
Order Info
Shipping Condition: Room Temperature
Specifications
Store at room temperature and protect from light. | |
Fluorescence | |
Phosphoprotein Gel Stain | |
Room Temperature |
In-Gel Detection | |
Pro-Q Diamond | |
PRO-Q™ | |
Proteins (Phosphoproteins) |