missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PeppermintStick™ Phosphoprotein Molecular Weight Standards
Molecular weight range from 14,400 to 116,250
£217.00 - £282.00
Specifications
Product Type | Phosphoprotein Molecular Weight Standard |
---|---|
System Type | SDS-PAGE |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10267432
|
Invitrogen™
P27167 |
40 μL |
£217.00
Each |
Estimated Shipment: 29-04-2024
Log in to see stock. |
|||||
10358242
|
Invitrogen™
P33350 |
400 μL |
£282.00
Each |
Estimated Shipment: 07-05-2024
Log in to see stock. |
|||||
Description
PeppermintStick™ phosphoprotein molecular weight standards are a mixture of phosphorylated and nonphosphorylated proteins. Separation by polyacrylamide gel electrophoresis resolves the standard mixture into two phosphorylated and four nonphosphorylated protein bands.
- System: SDS-PAGE
- Stain Type: Unstained
- Ready to Load: No
- Gel Compatibility: NuPAGE™ Tris-Acetate Gels, Novex™Tricine Gels, NuPAGE™ Bis-Tris Gels, Novex™Tris-Glycine Gels, SDS-PAGE Gels
1D Gel Electrophoresis, Protein Gel Electrophoresis, Protein Gel Staining and Imaging, Proteins, Expression, Isolation and Analysis
Order Info
Shipping Condition: Room Temperature
Specifications
Phosphoprotein Molecular Weight Standard | |
SDS-PAGE |
For Research Use Only. Not for use in diagnostic procedures.