Learn More
Invitrogen™ LI Silver (LIS) Enhancement Kit
Convenient, light-insensitive silver enhancement system for use with colloidal gold and NANOGOLD reagents
Brand: Invitrogen™ L24919
168.67 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Code : 50
Additional Details : Weight : 0.00450kg
Description
Gold particles in the presence of silver (I) ions and a reducing agent act as catalysts to reduce the silver (I) ions to metallic silver. The metallic silver is deposited onto the gold, enlarging the particles to between 30 and 100nm in diameter. Tissue or blots stained with colloidal gold are “developed” by this autometallographic procedure to give black staining that can be seen in the light microscope.
The LI Silver Enhancement Kit is prepared for Molecular Probes by Nanoprobes, Inc.
Protein Gel Electrophoresis, Protein Gel Staining and Imaging, Proteins, Expression, Isolation and Analysis, Western Blotting
Order Info
Shipping condition: Room temperature
Specifications
Store in refrigerator (2–8°C) and protect from light. | |
LIS Enhancement Kit | |
NANOGOLD™, FluoroNanogold™, Silver, Colloidal Gold | |
Room Temperature |
For Research Use Only. Not for use in diagnostic procedures.