missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FocalCheck™ Microspheres, 6 μm, fluorescent green ring stain/blue throughout
Fluorescent green/orange/dark-red ring stains
Brand: Invitrogen™ F14808
Code : 50
Additional Details : Weight : 0.00450kg
Description
The sharp ring stains exhibited by the 6.0 μm FocalCheck™ fluorescent green ring stained/blue throughout microspheres produce a striking visual representation of instrument misalignment or other aberrations making them ideal as reference standards for confocal laser-scanning microscopy. Correct image registration is indicated when the ring image and disk image of the combination beads are perfectly coincident in all dimensions.Specifications
Microsphere | |
Confocal Microscope Calibration | |
Suspension Beads | |
Store in refrigerator 2°C to 8°C and protect from light. | |
Room Temperature |
FocalCheck | |
Blue, Green | |
6 μm | |
1 Ea. |
For Research Use Only. Not for use in diagnostic procedures.