missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CoverWell™ Perfusion Chamber Gasket, 8 chambers, 9 mm diameter, 0.5 mm deep

Designed to facilitate live-cell imaging and manipulation

Brand:  Invitrogen™ C18121

Code : 50

Additional Details : Weight : 0.00450kg

 View more versions of this product

Product Code. 10003442

  • £199.00 / Each


Estimated Shipment date
from Supplier 23-04-2020
Add to basket

For Research Use Only. All usage must comply with product instructions.
 

Description

Description

  • CoverWell perfusion chamber gaskets form watertight “press-to-seal” chambers with dual-access ports so reagents can be quickly added and removed
  • Available in single- or multi-well configurations, these gaskets allow multiple experiments to be performed on single microscope slide or coverslip
  • Heat-resistant gaskets can be sterilized and used for direct culturing of cells and tissues

Applications:

Cell Analysis, Cellular Imaging, IHC Sample Preparation, IHC Staining & Detection, Immunocytochemistry (ICC), Immunofluorescence (IF), Immunofluorescence Staining & Detection, Immunohistochemistry (IHC)

Specifications

Specifications

Perfusion Chamber Gasket (1 chamber)
Facilitate live-cell imaging and manipulation
33 x 19mm
1.0mm
Videos
Documents

For Research Use Only. Not for use in diagnostic procedures.