Learn More
Invitrogen CoverWell™ imaging chamber gasket with adhesive, one chamber, 20 mm diameter, 0.5 mm deep Invitrogen
Brand:  Invitrogen C24726
CoverWell™ imaging chamber gasket with adhesive, one chamber, 20 mm diameter, 0.5 mm deep Invitrogen
Code : 50
Additional Details : Weight : 0.00450kg
Product Code. 10464972