Learn More
Invitrogen™ Molecular Probes™ CellLight™ Tubulin-GFP, BacMam 2.0
Provides easy method for labeling of tubulin with Green Fluorescent Protein (GFP) in live cells
Brand:  Invitrogen™ C10509
Code : NEW
Additional Details : Weight : 0.00450kg
 View more versions of this product
Product Code. 13296509
  • £84.67 / 100┬ÁL
EU Stock 7
Add to basket
Disclaimers:
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Specifications
100μL
488/510
Attune™ Acoustic Focusing Cytometer, Confocal Microscope, Floid™ Cell Imaging System, Fluorescence Microscope, Microscope
Green
Tubulin
GFP (EmGFP)
Fluorescent
Videos