Learn More
Invitrogen™ CellLight™ Lysosomes-GFP, BacMam 2.0
Provides easy method for labeling of lysosomes with Green Fluorescent Protein in live cells
£108.00 - £576.00
Specifications
Color | Green |
---|---|
Content And Storage | Store at 2°C to 6°C, protected from light. Do Not Freeze. |
Excitation Wavelength Range | 488⁄510 |
Dye Type | GFP (EmGFP) |
Detection Method | Fluorescent |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10304759
|
Invitrogen™
C10596 |
1 vial |
£576.00
Each |
Estimated Shipment: 09-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12616197
|
Invitrogen™
C10507 |
100μL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
This ready-to-use construct is transduced/transfected into cells using BacMam 2.0 technology, where it expresses GFP fused to Lamp1 (lysosomal associated membrane protein 1). You can observe lysosomes-GFP behavior in live cells using a fluorescent imaging system and standard FITC/GFP filter set. Study staining of lysosomes, which does not depend upon organelle pH or some other parameter, that can be different from one cell type to other. In addition, you can co-transduce/transfect more than one CellLight reagent and label live cells with multiple tracking or tracing dyes to image dynamic cellular processes. Cells expressing CellLight constructs can also be fixed with formaldehyde for multiplexing with antibodies using immunocytochemical techniques.
BacMam Technology
CellLight Lysosomes-GFP, BacMam 2.0, is a fusion construct of Lamp1 (lysosomal associated membrane protein 1) and emGFP, providing accurate and specific targeting to cellular lysosomes-GFP. This fusion construct is packaged in the insect virus baculovirus, which does not replicate in human cells and is designated as safe to use with biosafety level (BSL) 1 in most laboratories. BacMam technology ensures that most mammalian cell types are transduced/transfected with high efficiency and minimal toxicity. This transient transfection can be detected after overnight incubation for up to five days—enough time to carry out most dynamic cellular analyses. Like any transfection/transduction technique, the BacMam method does not transfect/transduce all of the cells with equal efficiency, making it poorly suited to cellular population studies or automated imaging/counting. CellLight reagents are ideal for experiments where cellular or subcellular co-locatization is required, or for cellular function studies that need special resolution.
- Highly efficient: up to 90% transduction of wide range of mammalian cell lines, including primary cells, stem cells, and neurons
- Fast and convenient: simply add CellLight reagent to cells, incubate overnight, and image—or store frozen, assay-ready cells for later use
- Less toxic: CellLight reagents are non-replicating in mammalian cells and are suitable for biosafety level (BSL) 1 handling
- Flexible: co-transduce more than one BacMam reagent for multiplex experiments or co-localization studies; tightly control expression levels by simply varying the dose
- Flow Cytometer Laser Lines: 488
Cell Analysis, Cell Structure, Cell Viability, Proliferation & Function, Cellular Imaging, Chemotaxis & Cell Migration, Cytoskeleton, Immunocytochemistry (ICC), Immunofluorescence (IF), Immunofluorescence Staining & Detection
Order Info
Shipping Condition: Wet Ice
Specifications
Green | |
488⁄510 | |
Fluorescent | |
Liquid | |
Wet Ice | |
Lysosomal Probe |
Store at 2°C to 6°C, protected from light. Do Not Freeze. | |
GFP (EmGFP) | |
Visible | |
CellLight™ | |
Fluorescent Dye | |
Lysosomes |
For Research Use Only. Not for use in diagnostic procedures.