missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Molecular BioProducts™ PCR Tubes and Plates

Molded from 100% virgin polypropylene

Brand:  Thermo Scientific™ 3421-11

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 12338202

  • £90.41 / Pack of 25


Call for latest availability
+44 (0)1509 555 500
Add to basket

Description

Description

Tubes feature thin walls for optimal heat transfer and caps that seal securely to minimize evaporation. Plates feature elevated well rims to ensure uniform sealing.

  • PurePak™ packaging reduces contamination from reaching into a bulk bag of tubes by dividing tubes into 10 separate PurePaks
  • Certified RNase-, DNase-, DNA, and Pyrogen-free
  • Clear, nonsterile

Specifications

Specifications

32-well PCR Plates
Natural
0.2mL
NA
0.2mL
Videos