missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Molecular Biological Water, Waternation
Click to view available options
Quantity:
10 x 100 mL
10 x 500 mL
6 x 1000 mL
Description
- Ready to use
- No chemical additives
- Batch traceability
- Certificate of conformity and analysis
- Reduce expensive test fails due to impurities
- High performance
- High accuracy
- Suppress water-forming side reactions
- Provide consistent performance across many sample types
The MLG range, a comprehensive suite of five high grade pure waters, in assorted sizes, ranging from simple deionised to molecular biology grades, catering to the majority of pure water lab uses.
Specifications
Specifications
pH | 7 |
Conductivity | 0.1 μS/cm |
Grade | Molecular Biology |
Packaging | PET Plastic Bottle |
Certificate of Analysis | Yes |
Quantity | 10 x 500 mL |
For Use With (Application) | Antibody Testing |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'