Learn More
Moldex™ 8000 Series Half-Mask Facepieces
Soft, durable facepiece confroms to the contours of the face without pressure points to provide long lasting comfort
Brand: Moldex™ 800201
Code : S8
Additional Details : Weight : 0.17108kg
Description
Soft, durable facepiece confroms to the contours of the face without pressure points to provide long lasting comfort
- Extra wide surface seals easily and stays sealed even when talking
- Low profile design provides a large field of vision
- Facepiece has no gaskets or yoke; fewer parts to maintain and inventory
- Each cartridge includes a new inhalation valve and features snap-in mount that eliminates mis-threading
Specifications
NIOSH | |
Thermoplastic Elastomer | |
Medium |
Snap-in gasket | |
Half Mask Facepiece |