Learn More
DuPont™ Model CHZ5 Tychem™ F2 Hooded Coverall
Brand:  DuPont™ D13489146
Code : NEW
Additional Details : Weight : 0.01000kg
Product Code. 12613660
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »
Specifications
Specifications
Zipper, Flap
Tychem™ Fabric
Elastic
Double
Gray
Elastic
Elastic
Hooded Coverall
Tychem F2 Fabric
124 to 132cm
Hooded
StitchedþTaped
3X-Large
for splash or pressurized splash protection in a variety of demanding industrial environments, including pulp and paper manufacturing, shutdowns, chemical processing and pharmaceutical manufacturing. It can be used across a broad range of applications, from chemical spill clean-up, emergency response, to military and petrochemical applications
Category III, Type 3-B, 4-B, 5-B and 6-B
Videos