Learn More
DuPont™ Model CHZ5 Tychem™ C2 Coverall
Catalog Number 12673650
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
3X-Large
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Type Hooded Coveralls
Size Large
Material Tychem
Color Yellow
Waist Style Elastic
Ankle Style Elastic
Cuff Style Double
Neck Style Hooded
Hood Type Elastic
Closure Type Zipper with Storm Flap
Seam Type Stitched, Taped
Protection Class Category III, Type 3-B, 4-B, 5-B and 6-B
Description Tychem C2 Fabric
Size (Metric) Chest 100 to 108 cm
For Use With (Application) For splash or pressurized splash protection in a variety of demanding industrial environments, including pulp and paper manufacturing, shutdowns, chemical processing and pharmaceutical manufacturing