missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DuPont™ Tyve 600 Plus Green Hooded Coverall
787.89 GBP valid until 2024-12-31
Use promo code "21990" to get your promotional price.
748.50 GBP valid until 2024-04-30
Use promo code "21673" to get your promotional price.
Use promo code "21990" to get your promotional price.
748.50 GBP valid until 2024-04-30
Use promo code "21673" to get your promotional price.
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
Description
- Self-adhesive chin strap for tight seal of suit to face mask
- Stitched and over-taped seams for protection and strength
- Tyvek zipper and zipper flap for enhanced protection
- Elasticated face, wrists and ankles as well as a glued-in elastic waist
- Elasticated thumb loops prevent sleeves from riding up
- Permeable to both air and water vapour, yet repels water-based liquids and aerosols
- Excellent barrier against fine particles and fibres (down to 1 micron in size), is ultra-low-linting and antistatically treated
- Applications include maintenance and dismantling jobs in the nuclear industry, pharmaceutical manufacturing or research, biosecurity laboratories as well as in medical applications and when exposed to biological hazards
Specifications
Product Type | Hooded Coveralls |
Size | 2X-Large |
Material | Tyvek |
Color | Green |
Waist Style | Elastic |
Ankle Style | Elastic |
Cuff Style | Elastic |
Neck Style | Hooded |
Hood Type | Elastic |
Closure Type | Zipper with Storm Flap |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'