Learn More
DuPont™ Tyve 600 Plus Green Hooded Coverall
Providing an ideal balance of protection, durability and comfort. DuPont™ Tyvek™ 600 Plus Green Hooded Coveralls are composed of flash spun high density polyethylene.
Brand: DuPont™ D13495738
Code : 94
Additional Details : Weight : 10.80000kg
Description
- Self-adhesive chin strap for tight seal of suit to face mask
- Stitched and over-taped seams for protection and strength
- Tyvek zipper and zipper flap for enhanced protection
- Elasticated face, wrists and ankles as well as a glued-in elastic waist
- Elasticated thumb loops prevent sleeves from riding up
- Permeable to both air and water vapour, yet repels water-based liquids and aerosols
- Excellent barrier against fine particles and fibres (down to 1 micron in size), is ultra-low-linting and antistatically treated
- Applications include maintenance and dismantling jobs in the nuclear industry, pharmaceutical manufacturing or research, biosecurity laboratories as well as in medical applications and when exposed to biological hazards
Specifications
Elastic | |
Tyvek™ | |
Elastic | |
Self-adhesive chin flap | |
Elastic | |
Hooded | |
X-Large | |
<250 g |
Zipper with storm flap | |
Category III, Type 4-B, 5-B and 6-B | |
(EU) 2016/425, EN 14126 | |
Green | |
Applications for Tyvek™ Classic Plus garments include maintenance and dismantling jobs in the nuclear industry, pharmaceutical manufacturing or in research and biosecurity laboratories, as well as in medical applications and when exposed to biological hazards | |
Stitched, Taped | |
108 to 116 cm |