missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DuPont™ Tychem™ 6000 F Orange Hooded Coverall
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
Description
- Available in grey for discretionary needs
- Elasticated face, wrists, waist and ankles for optimal fit
- Thumb loops prevent sleeves from riding up
- Self-adhesive chin flap for tight seal of suit to face masks
- Self-adhesive double zipper flap closure system
- Stitched and over-taped seams with barrier tape for protection and strength
- Protects personnel against numerous toxic industrial organic chemicals, highly concentrated inorganic chemicals (even under pressure), particulates, radioactive contamination, biohazards and certain chemical warfare agents
- Used for a broad range of applications from chemical spill clean-up, emergency response to military and petrochemical applications
Specifications
Product Type | Hooded Coveralls |
Size | Large |
Material | Tychem |
Color | Orange |
Waist Style | Elastic |
Ankle Style | Elastic |
Cuff Style | Elastic |
Neck Style | Hooded |
Hood Type | Elastic |
Closure Type | Double Zipper with Self-adhesive Flap |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'