Learn More
DuPont™ Tychem 6000 F CHA5 Hooded Coverall
Offering excellent chemical permeation protection to an extensive range of chemicals. DuPont™ Tychem™ 6000 F Hooded Coveralls are made from a lightweight and durable fabric consisting of a proprietary barrier film-laminated to a heavy duty Tyvek substrate.
Brand: DuPont™ D13495186
Code : NEW
Additional Details : Weight : 0.01000kg
Description
- Available in grey for discretionary needs
- Elasticated face, wrists, waist and ankles for optimal fit
- Thumb loops prevent sleeves from riding up
- Self-adhesive chin flap for tight seal of suit to face masks
- Self-adhesive double zipper flap closure system
- Stitched and over-taped seams with barrier tape for protection and strength
- Protects personnel against numerous toxic industrial organic chemicals, highly concentrated inorganic chemicals (even under pressure), particulates, radioactive contamination, biohazards and certain chemical warfare agents
- Used for a broad range of applications from chemical spill clean-up, emergency response to military and petrochemical applications
Specifications
Hooded Coveralls | |
Tychem | |
Elastic | |
Elastic | |
Elastic | |
Stitched, Taped | |
Chemicals | |
(EU) 2016/425, EN 14126, EN 1073-2, EN 1149-5 | |
For a broad range of applications from chemical spill clean-up, emergency response, to military and petrochemical applications |
Small | |
Gray | |
Elastic | |
Hooded | |
Double Zipper with Self-adhesive Flap | |
Category III, Type 3-B, 4-B, 5-B and 6-B | |
Self-adhesive chin flap | |
84 to 92 cm |