Learn More
Moclobemide, Tocris Bioscience™
Reversible MAO-A inhibitor
Brand:  RD Systems (Bio-Techne) 4395/50
Code : Z1
Additional Details : Weight : 0.00050kg
Quantity 50mg
 View more versions of this product
Product Code. 13454684
  • £417.48 / 50mg
Estimated Shipment date
from Supplier 01-04-2020
Description
Description
Reversible monoamine oxidase A (MOA-A) inhibitor.
Specifications
Chemical Identifiers
71320-77-9
268.741
moclobemide, aurorix, manerix, moclamine, moclobemid, moclobemida, moclobemidum, moclaime, moclamide, moclobamide
CHEBI:83531
C1COCCN1CCNC(=O)C2=CC=C(C=C2)Cl
C13H17ClN2O2
YHXISWVBGDMDLQ-UHFFFAOYSA-N
4235
4-chloro-N-(2-morpholin-4-ylethyl)benzamide
Specifications
50mg
71320-77-9
moclobemide, aurorix, manerix, moclamine, moclobemid, moclobemida, moclobemidum, moclaime, moclamide, moclobamide
C1COCCN1CCNC(=O)C2=CC=C(C=C2)Cl
268.741
CHEBI:83531
>99%
Moclobemide
C13H17ClN2O2
YHXISWVBGDMDLQ-UHFFFAOYSA-N
4-chloro-N-(2-morpholin-4-ylethyl)benzamide
4235
268.74