missing translation for 'onlineSavingsMsg'
Learn More

MNI-caged-D-aspartate, Tocris Bioscience™

Caged D-aspartate; NMDA agonist

Brand:  Tocris (Bio-Techne) 227710

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 10mg

 View more versions of this product

Product Code. 13440924

  • £170.52 / 10mg

Estimated Shipment date
from Supplier 23-10-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

D-aspartate caged with the photosensitive 4-methoxy-7-nitroindolinyl group. Photolyzed by UV light with a quantum efficiency of 0.09 at pH 7.4. Agonist at NMDA receptors and EAAT substrate.
Specifications

Chemical Identifiers

845555-94-4
309.278
mni-caged-d-aspartate, 4-methoxy-7-nitroindolinyl-caged-d-aspartate, alphar-alpha-amino-2,3-dihydro-4-methoxy-7-nitro-gamma-oxo-1h-indole-1-butanoic acid, alphar-alpha-amino-gamma-oxo-4-methoxy-7-nitro-2,3-dihydro-1h-indole-1-butyric acid, r-?-amino-2,3-dihydro-4-methoxy-7-nitro-?-oxo-1h-indole-1-butanoic acid
(2R)-2-amino-4-(4-methoxy-7-nitro-2,3-dihydroindol-1-yl)-4-oxobutanoic acid
C13H15N3O6
ATNVWKDWOLQDKH-MRVPVSSYSA-N
56972233
COC1=C2CCN(C2=C(C=C1)[N+](=O)[O-])C(=O)CC(C(=O)O)N

Specifications

10mg
845555-94-4
mni-caged-d-aspartate, 4-methoxy-7-nitroindolinyl-caged-d-aspartate, alphar-alpha-amino-2,3-dihydro-4-methoxy-7-nitro-gamma-oxo-1h-indole-1-butanoic acid, alphar-alpha-amino-gamma-oxo-4-methoxy-7-nitro-2,3-dihydro-1h-indole-1-butyric acid, r-?-amino-2,3-dihydro-4-methoxy-7-nitro-?-oxo-1h-indole-1-butanoic acid
COC1=C2CCN(C2=C(C=C1)[N+](=O)[O-])C(=O)CC(C(=O)O)N
309.278
309.27
MNI-caged-D-aspartate
C13H15N3O6
ATNVWKDWOLQDKH-MRVPVSSYSA-N
(2R)-2-amino-4-(4-methoxy-7-nitro-2,3-dihydroindol-1-yl)-4-oxobutanoic acid
56972233
>98%
Videos
Documents