Learn More
Tocris Bioscience™ MMPX
Brand:  Tocris Bioscience™ 055250
Code : Z1
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 13447753
  • £654.15 / 50mg
Call for latest availability
+44 (0)1509 555 500
Description
Description
Specific inhibitor of calmodulin-sensitive cyclic GMP phosphodiesterase (IC50 = 5.2 μM).
Specifications
Specifications
8-Methoxymethyl-3-isobutyl-1-methylxanthine
C12H18N4O3
50mg
78033-08-6
Observed MW: 266.3
Soluble to 100 mM in ethanol