Learn More
Tocris Bioscience™ MMPX
Brand:  Tocris Bioscience™ 055210
Code : Z1
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 13437753
  • £151.80 / 10mg
Estimated Shipment date
from Supplier 24-09-2019
Description
Description
Specific inhibitor of calmodulin-sensitive cyclic GMP phosphodiesterase (IC50 = 5.2 μM).
Specifications
Specifications
8-Methoxymethyl-3-isobutyl-1-methylxanthine
C12H18N4O3
10mg
78033-08-6
Observed MW: 266.3
Soluble to 100 mM in ethanol