missing translation for 'onlineSavingsMsg'
Learn More

MMPIP hydrochloride, Tocris Bioscience™

Potent, allosteric mGlu7-selective antagonist

Brand:  RD Systems (Bio-Techne) 29631

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 1mg

 View more versions of this product

Product Code. 13462144

  • £90.45 / 1mg


Estimated Shipment
28-02-2019
Add to basket

Description

Description

Potent allosteric mGlu7-selective receptor antagonist. Inhibits agonist-induced intracellular calcium mobilization and cAMP accumulation (IC50 values are 26 and 610 nM). Displays intrinsic activity; increases forskolin-induced cAMP accumulation in the absence of an agonist (IC50 = 15 nM), indicating inverse agonist activity.
Specifications

Chemical Identifiers

479077-02-6
333.347
mmpip, mmpip hydrochloride, 6-4-methoxyphenyl-5-methyl-3-pyridin-4-yl isoxazolo 4,5-c pyridin-4 5h-one, 6-4-methoxyphenyl-5-methyl-3-pyridin-4-yl-1,2 oxazolo 4,5-c pyridin-4-one, d08uow, mmpip hplc, 6-4-methoxyphenyl-5-methyl-3-4-pyridinyl-isoxazolo 4,5-c pyridin-4 5h-one, 6-4-methoxyphenyl-5-methyl-3-4-pyridinyl-isoxazolo 4,5-c pyridin-4 5h-one, 6-4-methoxyphenyl-5-methyl-3-4-pyridyl isoxazolo 4,5-c pyridine-4 5h-one, 6-4-methoxyphenyl-5-methyl-3-pyridin-4-yl-4h,5h-1,2 oxazolo 4,5-c pyridin-4-one
6-(4-methoxyphenyl)-5-methyl-3-pyridin-4-yl-[1,2]oxazolo[4,5-c]pyridin-4-one
C19H15N3O3
PDWYBOZNEVALOV-UHFFFAOYSA-N
9945530
CN1C(=CC2=C(C1=O)C(=NO2)C3=CC=NC=C3)C4=CC=C(C=C4)OC

Specifications

MMPIP hydrochloride
479077-02-6
mmpip, mmpip hydrochloride, 6-4-methoxyphenyl-5-methyl-3-pyridin-4-yl isoxazolo 4,5-c pyridin-4 5h-one, 6-4-methoxyphenyl-5-methyl-3-pyridin-4-yl-1,2 oxazolo 4,5-c pyridin-4-one, d08uow, mmpip hplc, 6-4-methoxyphenyl-5-methyl-3-4-pyridinyl-isoxazolo 4,5-c pyridin-4 5h-one, 6-4-methoxyphenyl-5-methyl-3-4-pyridinyl-isoxazolo 4,5-c pyridin-4 5h-one, 6-4-methoxyphenyl-5-methyl-3-4-pyridyl isoxazolo 4,5-c pyridine-4 5h-one, 6-4-methoxyphenyl-5-methyl-3-pyridin-4-yl-4h,5h-1,2 oxazolo 4,5-c pyridin-4-one
CN1C(=CC2=C(C1=O)C(=NO2)C3=CC=NC=C3)C4=CC=C(C=C4)OC
333.347
369.8
1mg
C19H15N3O3
PDWYBOZNEVALOV-UHFFFAOYSA-N
6-(4-methoxyphenyl)-5-methyl-3-pyridin-4-yl-[1,2]oxazolo[4,5-c]pyridin-4-one
9945530
>98%
Videos