missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse MMP-3 Antibody, R&D Systems™
Goat Polyclonal Antibody has been used in 1 publication
POA
Specifications
Antigen | MMP-3 |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL |
Applications | Western Blot, Immunoprecipitation |
Classification | Polyclonal |
Description
MMP-3 Polyclonal specifically detects MMP-3 in Mouse samples. It is validated for Western Blot, Immunoprecipitation.Specifications
MMP-3 | |
Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL | |
Polyclonal | |
Goat | |
Mouse | |
CHDS6, EC 3.4.24, EC 3.4.24.17, matrix metallopeptidase 3 (stromelysin 1, progelatinase), matrix metalloproteinase 3 (stromelysin 1, progelatinase), Matrix metalloproteinase-3, MGC126102, MGC126103, MMP3, MMP-3, proteoglycanase, SL-1, STMY, STMY1MGC126104, STR1, Stromelysin 1, stromelysin-1, Transin-1 | |
MMP3 | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Western Blot, Immunoprecipitation | |
Unconjugated | |
RUO | |
P28862 | |
4314 | |
Mouse myeloma cell line NS0-derived recombinant mouse MMP-3 Tyr18-Cys477 Accession # P28862 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects mouse MMP-3 in direct ELISAs and Western blots. In direct ELISAs, approximately 50% cross-reactivity with recombinant human MMP-3 is observed and less than 1% cross-reactivity with recombinant mouse MMP-2 is observed. |
For Research Use Only