missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MMP-11 Mouse anti-Human, Clone: 135421, R&D Systems™
Mouse Monoclonal Antibody has been used in 1 publication
POA
Specifications
Antigen | MMP-11 |
---|---|
Clone | 135421 |
Immunogen | E. coli-derived recombinant human MMP-11 Met1-Leu488 Accession # P24347 |
Host Species | Mouse |
Primary or Secondary | Primary |
Description
MMP-11 Monoclonal specifically detects MMP-11 in Human samples. It is validated for Intracellular Staining by Flow Cytometry, CyTOF-ready.Specifications
MMP-11 | |
E. coli-derived recombinant human MMP-11 Met1-Leu488 Accession # P24347 | |
Primary | |
Human | |
Unconjugated | |
4320 | |
Purified | |
Flow Cytometry, CyTOF | |
RUO |
135421 | |
Mouse | |
Monoclonal | |
IgG2b | |
EC 3.4.24, EC 3.4.24.-, EC 3.4.24.24, matrix metallopeptidase 11 (stromelysin 3), matrix metalloproteinase 11 (stromelysin 3), Matrix metalloproteinase-11, MMP11, MMP-11, SL-3, ST3, STMY3stromelysin 3, stromelysin III, stromelysin-3 | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only