missing translation for 'onlineSavingsMsg'
Learn More

MM 77 dihydrochloride, Tocris Bioscience™

5-HT1A (postsynaptic) antagonist

Brand:  Tocris (Bio-Techne) 093310

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 10mg

 View more versions of this product

Product Code. 13458173

  • £92.61 / 10mg

Estimated Shipment date
from Supplier 30-03-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

A highly potent and fairly selective 5-HT1A ligand, which may be a full antagonist at postsynaptic receptors.
Specifications

Chemical Identifiers

159187-70-9
418.359
mm 77 dihydrochloride, 1-2-methoxyphenyl-4-4-succinimidobutyl piperazine dihydrochloride, 1-4-4-2-methoxyphenyl piperazin-1-yl butyl pyrrolidine-2,5-dione dihydrochloride, 1-2-methoxyphenyl-4-4-succinimidobutyl piperazinedihydrochloride, 2,5-pyrrolidinedione,1-4-4-2-methoxyphenyl-1-piperazinyl butyl, 1-4-4-2-methoxyphenyl piperazin-1-yl butyl pyrrolidine-2,5-dione-hydrogen chloride 1/2
1-[4-[4-(2-methoxyphenyl)piperazin-1-yl]butyl]pyrrolidine-2,5-dione;dihydrochloride
C19H29Cl2N3O3
ZYCVVOZXHNDRCO-UHFFFAOYSA-N
45073441
COC1=CC=CC=C1N2CCN(CC2)CCCCN3C(=O)CCC3=O.Cl.Cl

Specifications

10mg
159187-70-9
mm 77 dihydrochloride, 1-2-methoxyphenyl-4-4-succinimidobutyl piperazine dihydrochloride, 1-4-4-2-methoxyphenyl piperazin-1-yl butyl pyrrolidine-2,5-dione dihydrochloride, 1-2-methoxyphenyl-4-4-succinimidobutyl piperazinedihydrochloride, 2,5-pyrrolidinedione,1-4-4-2-methoxyphenyl-1-piperazinyl butyl, 1-4-4-2-methoxyphenyl piperazin-1-yl butyl pyrrolidine-2,5-dione-hydrogen chloride 1/2
COC1=CC=CC=C1N2CCN(CC2)CCCCN3C(=O)CCC3=O.Cl.Cl
418.359
418.36
MM 77 dihydrochloride
C19H29Cl2N3O3
ZYCVVOZXHNDRCO-UHFFFAOYSA-N
1-[4-[4-(2-methoxyphenyl)piperazin-1-yl]butyl]pyrrolidine-2,5-dione;dihydrochloride
45073441
>98%
Videos
Documents