Learn More
MLF2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA552745
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: MFRFMRDVEP EDPMFLMDPF AIHRQHMSRM LSGGFGYSPF LSITDGNMPG TRPASRRMQQ A Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 95%.
MLF2 Polyclonal antibody specifically detects MLF2 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin).Specifications
MLF2 | |
Polyclonal | |
Unconjugated | |
MLF2 | |
AI256856; AU018944; C77284; MLF2; Myelodysplasia-myeloid leukemia factor 2; myeloid leukemia factor 2; myeloid leukemia factor 2 homolog; NTN4 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
8079 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin) | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q15773 | |
MLF2 | |
Recombinant protein corresponding to Human MLF2 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only