missing translation for 'onlineSavingsMsg'
Learn More

ML 9 hydrochloride, Tocris Bioscience™

Myosin light chain kinase inhibitor

Brand:  Tocris (Bio-Techne) 043150

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 50mg

 View more versions of this product

Product Code. 13467643

  • £590.94 / 50mg

Estimated Shipment date
from Supplier 30-03-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Selective MLCK inhibitor (Ki values are 4, 32 and 54 μM for MLCK, PKA and PKC respectively). Also inhibits STIM1-plasma membrane interactions, preventing store-operated Ca2+ entry (SOCE).
Specifications

Chemical Identifiers

105637-50-1
361.281
ml-9, ml 9, ml-9, hydrochloride, unii-bz2l6jk93i, ml 9 hydrochloride, 1-5-chloronaphthalene-1-sulfonyl-1h-hexahydro-1,4-diazepine hcl, bz2l6jk93i, 1-5-chloronaphthalenesulfonyl homopiperazine hydrochloride, 1-5-chloronaphthalene-1-sulfonyl-1h-hexahydro-1,4-diazepine, 1-5-chloronaphthalene-1-sulfonyl homopiperazine, hcl
1-(5-chloronaphthalen-1-yl)sulfonyl-1,4-diazepane;hydrochloride
C15H18Cl2N2O2S
ZNRYCIVTNLZOGI-UHFFFAOYSA-N
108047
C1CNCCN(C1)S(=O)(=O)C2=CC=CC3=C2C=CC=C3Cl.Cl

Specifications

50mg
105637-50-1
ml-9, ml 9, ml-9, hydrochloride, unii-bz2l6jk93i, ml 9 hydrochloride, 1-5-chloronaphthalene-1-sulfonyl-1h-hexahydro-1,4-diazepine hcl, bz2l6jk93i, 1-5-chloronaphthalenesulfonyl homopiperazine hydrochloride, 1-5-chloronaphthalene-1-sulfonyl-1h-hexahydro-1,4-diazepine, 1-5-chloronaphthalene-1-sulfonyl homopiperazine, hcl
C1CNCCN(C1)S(=O)(=O)C2=CC=CC3=C2C=CC=C3Cl.Cl
361.281
361.29
ML 9 hydrochloride
C15H18Cl2N2O2S
ZNRYCIVTNLZOGI-UHFFFAOYSA-N
1-(5-chloronaphthalen-1-yl)sulfonyl-1,4-diazepane;hydrochloride
108047
>99%
Videos
Documents