missing translation for 'onlineSavingsMsg'
Learn More

ML 213, Tocris Bioscience™

KV7.2 and KV7.4 channel opener

Brand:  RD Systems (Bio-Techne) 4519/10

Code : Z1

Additional Details : Weight : 0.00010kg

Quantity 10mg

 View more versions of this product

Product Code. 13434874

  • £173.46 / 10mg

Estimated Shipment date
from Supplier 20-11-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Selective KV7.2 (KCNQ2) and KV7.4 (KCNQ4) channel opener (EC50 values are 230 and 510 nM for KV7.2 and KV7.4 respectively). Displays > 80-fold selectivity against KV7.1, KV7.3 and KV7.5 in a thallium-based fluorescence assay. Hyperpolarizes resting membrane potential in an in vitro spinal cord preparation.
Specifications

Chemical Identifiers

489402-47-3
257.377
n-mesitylbicyclo 2.2.1 heptane-2-carboxamide, n-2,4,6-trimethylphenyl-bicyclo 2.2.1 heptane-2-carboxamide, n-2,4,6-trimethylphenyl bicyclo 2.2.1 heptane-2-carboxamide, n-2,4,6-trimethylphenyl bicyclo 2.2.1 heptane-3-carboxamide, bicyclo 2.2.1 heptane-2-carboxylic acid 2,4,6-trimethyl-phenyl-amide, d0xy2m, qcr-50, regid_for_cid_3111211, bicyclo 2.2.1 hept-2-yl-n-2,4,6-trimethylphenyl carboxamide, n-2,4,6-trimethylphenyl bicyclo 2.2.1 heptane-6-carboxamide
N-(2,4,6-trimethylphenyl)bicyclo[2.2.1]heptane-3-carboxamide
C17H23NO
SIQGKPGBLYKQBB-UHFFFAOYSA-N
3111211
CC1=CC(=C(C(=C1)C)NC(=O)C2CC3CCC2C3)C

Specifications

10mg
489402-47-3
n-mesitylbicyclo 2.2.1 heptane-2-carboxamide, n-2,4,6-trimethylphenyl-bicyclo 2.2.1 heptane-2-carboxamide, n-2,4,6-trimethylphenyl bicyclo 2.2.1 heptane-2-carboxamide, n-2,4,6-trimethylphenyl bicyclo 2.2.1 heptane-3-carboxamide, bicyclo 2.2.1 heptane-2-carboxylic acid 2,4,6-trimethyl-phenyl-amide, d0xy2m, qcr-50, regid_for_cid_3111211, bicyclo 2.2.1 hept-2-yl-n-2,4,6-trimethylphenyl carboxamide, n-2,4,6-trimethylphenyl bicyclo 2.2.1 heptane-6-carboxamide
CC1=CC(=C(C(=C1)C)NC(=O)C2CC3CCC2C3)C
257.377
257.37
ML 213
C17H23NO
SIQGKPGBLYKQBB-UHFFFAOYSA-N
N-(2,4,6-trimethylphenyl)bicyclo[2.2.1]heptane-3-carboxamide
3111211
>99%
Videos
Documents