missing translation for 'onlineSavingsMsg'
Learn More

ML 145, Tocris Bioscience™

GPR35 antagonist

Brand:  Tocris (Bio-Techne) 417250

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 50mg

 View more versions of this product

Product Code. 13444284

  • £795.27 / 50mg

Estimated Shipment date
from Supplier 21-11-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Selective antagonist for the GPR35 orphan receptor GPCR (IC50 = 20.1 nM). Over 1000-fold more selective for GPR35 compared to GPR55 antagonists.
Specifications

Chemical Identifiers

1164500-72-4
482.569
2-hydroxy-4-4-5-2-methyl-3-phenyl-2-propen-1-ylidene-4-oxo-2-thioxo-1,3-thiazolidin-3-yl butanoyl amino benzoic acid, 2-hydroxy-4-4-5z-5-e-2-methyl-3-phenylprop-2-enylidene-4-oxo-2-sulfanylidene-1,3-thiazolidin-3-yl butanoylamino benzoic acid, 2-hydroxy-4-4-5z-5-2e-2-methyl-3-phenylprop-2-en-1-ylidene-4-oxo-2-thioxo-1,3-thiazolidin-3-yl butanoyl amino benzoic acid, 2-hydroxy-4-4-5z-5-e-2-methyl-3-phenylprop-2-enylidene-4-oxo-2-sulfanylidene-3-thiazolidinyl-1-oxobutyl amino benzoic acid, 2-hydroxy-4-4-5z-4-keto-5-e-2-methyl-3-phenyl-prop-2-enylidene-2-thioxo-thiazolidin-3-yl butanoylamino benzoic acid, 2-hydroxy-4-4-5z-5-2e-2-methyl-3-phenylprop-2-en-1-ylidene-4-oxo-2-sulfanylidene-1,3-thiazolidin-3-yl butanamido benzoic acid, 4-4-5z-5-e-2-methyl-3-phenyl-prop-2-enylidene-4-oxidanylidene-2-sulfanylidene-1,3-thiazolidin-3-yl butanoylamino-2-oxidanyl-benzoic acid
2-hydroxy-4-[4-[(5Z)-5-[(E)-2-methyl-3-phenylprop-2-enylidene]-4-oxo-2-sulfanylidene-1,3-thiazolidin-3-yl]butanoylamino]benzoic acid
C24H22N2O5S2
COFMYJWNXSFLKQ-QIROLCGISA-N
2286812
CC(=CC1=CC=CC=C1)C=C2C(=O)N(C(=S)S2)CCCC(=O)NC3=CC(=C(C=C3)C(=O)O)O

Specifications

50mg
1164500-72-4
2-hydroxy-4-4-5-2-methyl-3-phenyl-2-propen-1-ylidene-4-oxo-2-thioxo-1,3-thiazolidin-3-yl butanoyl amino benzoic acid, 2-hydroxy-4-4-5z-5-e-2-methyl-3-phenylprop-2-enylidene-4-oxo-2-sulfanylidene-1,3-thiazolidin-3-yl butanoylamino benzoic acid, 2-hydroxy-4-4-5z-5-2e-2-methyl-3-phenylprop-2-en-1-ylidene-4-oxo-2-thioxo-1,3-thiazolidin-3-yl butanoyl amino benzoic acid, 2-hydroxy-4-4-5z-5-e-2-methyl-3-phenylprop-2-enylidene-4-oxo-2-sulfanylidene-3-thiazolidinyl-1-oxobutyl amino benzoic acid, 2-hydroxy-4-4-5z-4-keto-5-e-2-methyl-3-phenyl-prop-2-enylidene-2-thioxo-thiazolidin-3-yl butanoylamino benzoic acid, 2-hydroxy-4-4-5z-5-2e-2-methyl-3-phenylprop-2-en-1-ylidene-4-oxo-2-sulfanylidene-1,3-thiazolidin-3-yl butanamido benzoic acid, 4-4-5z-5-e-2-methyl-3-phenyl-prop-2-enylidene-4-oxidanylidene-2-sulfanylidene-1,3-thiazolidin-3-yl butanoylamino-2-oxidanyl-benzoic acid
CC(=CC1=CC=CC=C1)C=C2C(=O)N(C(=S)S2)CCCC(=O)NC3=CC(=C(C=C3)C(=O)O)O
482.569
482.57
ML 145
C24H22N2O5S2
COFMYJWNXSFLKQ-QIROLCGISA-N
2-hydroxy-4-[4-[(5Z)-5-[(E)-2-methyl-3-phenylprop-2-enylidene]-4-oxo-2-sulfanylidene-1,3-thiazolidin-3-yl]butanoylamino]benzoic acid
2286812
>98%
Videos
Documents