missing translation for 'onlineSavingsMsg'
Learn More

ML 10302 hydrochloride, Tocris Bioscience™

Potent and selective 5-HT4 partial agonist

Brand:  Tocris (Bio-Techne) 349950

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 50mg

 View more versions of this product

Product Code. 13433084

  • £327.81 / 50mg

Estimated Shipment date
from Supplier 26-03-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Potent 5-HT4 partial agonist (EC50 = 4 nM) that displays > 680-fold selectivity over 5-HT3 receptors (Ki values are 1.07 and 730 nM respectively). Increases sAPPα levels in the cortex in an animal model of Alzheimer's disease and exhibits progastrokinetic effects in vivo.
Specifications

Chemical Identifiers

186826-17-5
349.252
ml 10302 hydrochloride, 4-amino-5-chloro-2-methoxybenzoic acid 2-1-piperidinyl ethyl ester hydrochloride, 2-piperidin-1-yl ethyl 4-amino-5-chloro-2-methoxybenzoate hydrochloride, 2-piperidin-1-yl ethyl 4-amino-5-chloro-2-methoxybenzoate-hydrogen chloride 1/1
2-piperidin-1-ylethyl 4-amino-5-chloro-2-methoxybenzoate;hydrochloride
C15H22Cl2N2O3
HBLRTSYSGOFQCK-UHFFFAOYSA-N
56972230
COC1=CC(=C(C=C1C(=O)OCCN2CCCCC2)Cl)N.Cl

Specifications

50mg
186826-17-5
ml 10302 hydrochloride, 4-amino-5-chloro-2-methoxybenzoic acid 2-1-piperidinyl ethyl ester hydrochloride, 2-piperidin-1-yl ethyl 4-amino-5-chloro-2-methoxybenzoate hydrochloride, 2-piperidin-1-yl ethyl 4-amino-5-chloro-2-methoxybenzoate-hydrogen chloride 1/1
COC1=CC(=C(C=C1C(=O)OCCN2CCCCC2)Cl)N.Cl
349.252
349.25
ML 10302 hydrochloride
C15H22Cl2N2O3
HBLRTSYSGOFQCK-UHFFFAOYSA-N
2-piperidin-1-ylethyl 4-amino-5-chloro-2-methoxybenzoate;hydrochloride
56972230
>99%
Videos
Documents