missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MKP2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA566440
Additional Details : Weight : 0.01000kg
Description
Immunogen sequence: QVLATSCAAEA ASPSGPLRER GKTPATPTSQ FVFSFPVSVG VHSAPSSLPY LHS Highest antigen sequence identity to the following orthologs - mouse 89%, rat 89%.
MKP2 Polyclonal antibody specifically detects MKP2 in Human samples. It is validated for Immunocytochemistry.Specifications
MKP2 | |
Polyclonal | |
Unconjugated | |
DUSP4 | |
2700078F24Rik; AI844617; BB104621; dual specificity phosphatase 4; dual specificity protein phosphatase 4; Dual specificity protein phosphatase hVH2; DUSP 4; DUSP4; DUSP-4; E130306H24Rik; HVH2; MAP kinase phosphatase 2; mitogen-activated protein kinase phosphatase 2; MKP II; Mkp2; Mkp-2; MKPII; serine/threonine specific protein phosphatase; TYP; VH1 homologous phosphatase 2; VH2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
1846 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q13115 | |
DUSP4 | |
Recombinant Human MKP2 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
For Research Use Only