Learn More
MKL1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA556557
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: EQEKRAQQPA PAPAPLGTPV KQENSFSSCQ LSQQPLGPAH PFNPSLAAPA TNHIDPCAVA PGPPSVVVKQ EALQPEPEPV PAPQLLLGPQ GPSLIKGVAP PTLITDSTGT HLVLTVTNKN ADSPG Highest antigen sequence identity to the following orthologs: Mouse - 65%, Rat - 66%.
The protein encoded by this gene interacts with the transcription factor myocardin, a key regulator of smooth muscle cell differentiation. The encoded protein is predominantly nuclear and may help transduce signals from the cytoskeleton to the nucleus. This gene is involved in a specific translocation event that creates a fusion of this gene and the RNA-binding motif protein-15 gene. This translocation has been associated with acute megakaryocytic leukemia.Specifications
MKL1 | |
Polyclonal | |
Unconjugated | |
Mrtfa | |
AMKL; basic SAP and coiled-coil domains; basic SAP coiled-coil transcription activator; basic, SAP and coiled-coil domain; basic, SAP, and coiled-coil domain containing protein; Bsac; FLMKL1; KIAA1438; Mal; megakaryoblastic leukemia (translocation) 1; megakaryoblastic leukemia (translocation) 1 homolog; megakaryoblastic leukemia 1 met; megakaryoblastic leukemia 1 protein; Megakaryoblastic leukemia 1 protein homolog; Megakaryocytic acute leukemia protein; Megakaryocytic acute leukemia protein homolog; MELODY; mixed lineage leukemia; MKL (megakaryoblastic leukemia)/myocardin-like 1; MKL/myocardin-like protein 1; MKL1; MKL1-elongated derivative of yield protein; MKL1met; MRTFA; MRTF-A; myocardin related transcription factor A; myocardin-related transcription factor A | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
57591 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
0.3 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q969V6 | |
Mrtfa | |
Recombinant protein corresponding to Human MKL1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only