Learn More
MK 212 hydrochloride, Tocris Bioscience™
5-HT2C agonist
Brand:  Tocris (Bio-Techne) 094150
Code : Z1
Additional Details : Weight : 0.01000kg
Quantity 50mg
 View more versions of this product
Product Code. 13448183
  • £366.03 / 50mg
Estimated Shipment date
from Supplier 19-11-2019
Description
Description
5-HT receptor agonist; displays selectivity for 5-HT2C over 5-HT2A (IC50 values are 0.028 and 0.42 μM for human 5-HT2C and 5-HT2A receptors expressed in HEK293 cells respectively).
Specifications
Chemical Identifiers
64022-27-1
198.654
2-chloro-6-1-piperazinyl pyrazine, 6-chloro-2-1-piperazinyl pyrazine, 2-chloro-6-piperazin-1-yl pyrazine, pyrazine, 2-chloro-6-1-piperazinyl, tocris-0941, biomol-nt_000130, d02esw, gtpl165, 1-6-chloro-2-pyrazinyl piperazine, 6-chloro-2-1-piperazinyl-pyrazine
2-chloro-6-piperazin-1-ylpyrazine
C8H11ClN4
CJAWPFJGFFNXQI-UHFFFAOYSA-N
107992
C1CN(CCN1)C2=CN=CC(=N2)Cl
Specifications
50mg
64022-27-1
2-chloro-6-1-piperazinyl pyrazine, 6-chloro-2-1-piperazinyl pyrazine, 2-chloro-6-piperazin-1-yl pyrazine, pyrazine, 2-chloro-6-1-piperazinyl, tocris-0941, biomol-nt_000130, d02esw, gtpl165, 1-6-chloro-2-pyrazinyl piperazine, 6-chloro-2-1-piperazinyl-pyrazine
C1CN(CCN1)C2=CN=CC(=N2)Cl
198.654
235.12
MK 212 hydrochloride
C8H11ClN4
CJAWPFJGFFNXQI-UHFFFAOYSA-N
2-chloro-6-piperazin-1-ylpyrazine
107992
>99%