missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mixed Multi-Element Calibration Standard 4A, SPEX CertiPrep™
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material
£135.00 - £228.00
Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
15266106
|
SPEX Certiprep
MXSTD4A-100N |
125 mL | Triple-leached LDPE bottle |
£135.00
125mL |
Estimated Shipment: 06-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
15276106
|
SPEX Certiprep
MXSTD4A-500N |
500 mL | Triple-leached LDPE bottle |
£228.00
500mL |
Estimated Shipment: 06-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Description
Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
Mixed Calibration StandardsCertifications
ISO/IEC 17025:2005, ISO 17034:2016, ISO 9001: 2015
Compliance
GHS/OSHA Complaint
Product intended for laboratory use only