missing translation for 'onlineSavingsMsg'
Learn More

Merck MF-Millipore™ Membrane Filter, Hydrophobic Edge, 3 μm

Brand:  Merck SSEP0470W

Code : 72

Additional Details : Weight : 0.01000kg Transport : UN number : 3270 Chem class : 4.1 Pack group : II

Product Code. 10710313

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Hydrophobic edge membranes are standard MF-Type filters with edges rendered hydrophobic to 3 or 6 mm widths. In antibiotic and bacteriostatic drug sterility testing by membrane filter method, the 6 mm wide non-wetting edge prevents drug intrusion under holder sealing rim.

Applications:

  • Microbiological and sterility testing of antibiotics and bacteriostatic drugs
  • Filling machine operations
  • Available in sterile or autoclave-ready packs
Specifications

Specifications

30 L/min/ cm2
White
MCE
Non-sterile
30 L/min/ cm2 (Air), 320 mL/min/ cm2 (water)
Hydrophobic Edge
100Pack
75°C
Filter Membranes
47 mm
≥0.7 bar
Membrane Filter
3 μm
SSEP
Hydrophobic edge membrane is a standard MF-Type filter with edges rendered hydrophobic to 6 mm widths
83%
Plain
150 μm
47 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers