missing translation for 'onlineSavingsMsg'
Learn More

Molecular Probes™ MitoProbe™ DiOC2(3) Assay Kit - For Flow Cytometry - 100 Assays

MitoProbe™ DiOC2(3) Assay Kit - for flow cytometry - 100 assays

Brand:  Molecular Probes™ M34150

Code : 50

Additional Details : Weight : 0.00450kg

Product Code. 10789613

  • £396.00 / Each
Estimated Shipment: 30-04-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


For Research Use Only. All usage must comply with product instructions.
 

Description

Description

The MitoProbe™ DiOC2(3) Assay Kit provides solutions of the membrane-potential-sensitive cyanine dye DiOC2(3), and a mitochondrial membrane-potential disrupter, CCCP, for the study of mitochondrial membrane potential. DiOC2(3) penetrates the cytosol of eukaryotic cells and accumulates primarily in mitochondria with active membrane potentials, producing bright, red fluorescence. DiOC2(3) staining intensity decreases with decreased mitochondrial membrane potential, accompanied by a shift from red to green fluorescence.

View a selection guide for all apoptosis assays for flow cytometry.
Specifications

Specifications

Contains 1 vial of DiOC2(3) (625 μL, 10 μM in DMSO), and 1 vial of CCCP: (125 μL, 50 mM DMSO). Store at 2–6°C, protected from light. Do not freeze.
Flow Cytometer
DiOC2(3)
Tube(s)
100
Fluorescent
Flow Cytometry Assay Kit
488
MitoProbe™
Room Temperature
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.