missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Misch Metal, 99.0% min. rare earth content, Thermo Scientific Chemicals
216.67 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
500 g
2 kg
Description
Misch Metal is used as dope for non-ferrous metal, such as aluminum (Al), iron-chrome-aluminum (Fe-Cr-Al), et al. This is to increase the operating life of Fe-Cr-Al heating devices. It is also used to manufacture flints in cigarette lighters.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Avg. Mol. Wt. or Mol. Wt. Range | Typically Ce:La 75:25 wt% |
UN Number | UN3178 |
Quantity | 500 g |
Solubility Information | Reacts with water. |
Percent Purity | ≥99.0% |
Sensitivity | Air and moisture sensitive |
Form | Pieces |
Assay | rare earth content |
Chemical Name or Material | Misch Metal |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'