missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse MIS/AMH Antibody, R&D Systems™
Rat Monoclonal Antibody
POA
Specifications
Antigen | MIS/AMH |
---|---|
Clone | 188103 |
Concentration | LYOPH |
Dilution | Western Blot 1 ug/mL |
Applications | Western Blot |
Description
MIS/AMH Monoclonal specifically detects MIS/AMH in Mouse samples. It is validated for Western Blot.Specifications
MIS/AMH | |
LYOPH | |
Western Blot | |
Unconjugated | |
Rat | |
Mouse | |
P27106.1 | |
268 | |
E. coli-derived recombinant mouse MIS/AMH Asp450-Arg555 Accession # P27106.1 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects mouse MIS/AMH in direct ELISAs and Western blots. In direct ELISAs and Western blots, this antibody does not cross-react with recombinant human MIS or recombinant rat MIS. |
188103 | |
Western Blot 1 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
AMH, MIF, MIS, Muellerian hormone, muellerian-inhibiting factor, muellerian-inhibiting substance, Mullerian hormone, Mullerian inhibiting factor, Mullerian inhibiting substance | |
AMH | |
IgG2b | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only