missing translation for 'onlineSavingsMsg'
Learn More

Mirin, Tocris Bioscience™

MRN-ATM pathway inhibitor

Brand:  Tocris (Bio-Techne) 319050

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 50mg

 View more versions of this product

Product Code. 13462394

  • £547.32 / 50mg


Estimated Shipment
28-02-2019
Add to basket

Description

Description

Mre11-Rad50-Nbs1 (MRN)-ATM pathway inhibitor that blocks the 3« and 5« exonuclease activity associated with Mre11. Prevents ATM activation in response to double strand breaks (IC50 = 12μM) and induces G2 cell cycle arrest. Also blocks homology-directed repair in vitro.
Specifications

Chemical Identifiers

1198097-97-0
220.246
mirin, z-5-4-hydroxybenzylidene-2-imino-1,3-thiazolidin-4-one, mrn-atm pathway inhibitor, mirin, 5z-5-4-hydroxyphenyl methylidene-2-imino-1,3-thiazolidin-4-one, 5-4-hydroxybenzylidene-2-iminothiazolidin-4-one, mrn-atm pathway inhibitor,mirin, mirin hplc , powder, 2-amino-5-4-hydroxyphenyl methylene-4 5h-thiazolone, 5z-5-4-hydroxybenzylidene-2-imino-1,3-thiazolidin-4-one, 5z-2-amino-5-4-hydroxyphenyl methylidene-1,3-thiazol-4-one
(5Z)-2-amino-5-[(4-hydroxyphenyl)methylidene]-1,3-thiazol-4-one
C10H8N2O2S
YBHQCJILTOVLHD-YVMONPNESA-N
1206243
C1=CC(=CC=C1C=C2C(=O)N=C(S2)N)O

Specifications

Mirin
1198097-97-0
mirin, z-5-4-hydroxybenzylidene-2-imino-1,3-thiazolidin-4-one, mrn-atm pathway inhibitor, mirin, 5z-5-4-hydroxyphenyl methylidene-2-imino-1,3-thiazolidin-4-one, 5-4-hydroxybenzylidene-2-iminothiazolidin-4-one, mrn-atm pathway inhibitor,mirin, mirin hplc , powder, 2-amino-5-4-hydroxyphenyl methylene-4 5h-thiazolone, 5z-5-4-hydroxybenzylidene-2-imino-1,3-thiazolidin-4-one, 5z-2-amino-5-4-hydroxyphenyl methylidene-1,3-thiazol-4-one
C1=CC(=CC=C1C=C2C(=O)N=C(S2)N)O
220.246
220.25
50mg
C10H8N2O2S
YBHQCJILTOVLHD-YVMONPNESA-N
(5Z)-2-amino-5-[(4-hydroxyphenyl)methylidene]-1,3-thiazol-4-one
1206243
>99%
Videos