missing translation for 'onlineSavingsMsg'
Learn More

Sartorius™ Minisart™ HY PTFE/ MBS Syringe Filters

Brand:  Sartorius™ 40078----------Q

UNSPSC : 41120000

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 11720836

  • £718.00 / Pack of 500


Call for latest availability
+44 (0)1509 555 500
Add to basket

Specifications Description & Specifications

Specifications

For Fast and Reliable Sterile Venting and Gas Filtration
MBS
Syringe Filter
Hose Barb
0.2μm
4.5Bar
Syringe Filter
Female Luer Lock
50°C
Clear