Learn More
Lovibond™ Water Testing Minikit
Lovibond™ Water Testing Minikit is easy to use and provides highly accurate results.
Brand: Lovibond™ 414270
Code : 04
Additional Details : Weight : 0.26000kg Transport : UN number : 3077 Chem class : 9 Pack group : III
Description
Lovibond™ Water Testing Minikit is easy to use and provides highly accurate results.
- Easy operation and exact reagent dosing
- High accuracy
- Foil-wrapped Lovibond™ tablet reagents with a minimum guaranteed shelf life of 5 years
- Unrestricted shipment
- Safe storage
Specifications
Nitrite | |
1 Pack | |
Approximately 40 Tests | |
Minikit water testing kit |
Flip top case containing a shaker tube, rapid dissolving tablet reagents and instructions. | |
70 to 1500mg/L | |
Tablet count |