Learn More
Merck Minicon B15, 8 Cells/Unit
Concentrate urine, cerebrospinal and other fluids to enrich analytes. Minicon B15, 8 Cells/Unit is used worldwide as a preparatory step to increase the sensitivity of subsequent in vitro and clinical tests. Static concentrator requires no accessories.
Brand: Merck 9031
Code : NEW
Additional Details : Weight : 1.08000kg
Description
Concentrate urine, cerebrospinal and other fluids to enrich analytes. Minicon B15, 8 Cells/Unit is used worldwide as a preparatory step to increase the sensitivity of subsequent in vitro and clinical tests. Static concentrator requires no accessories.
- Concentrate serum, urine, cerebrospinal fluid, and other body fluids prior to analysis
- Intesifies analytes that indicate abnormal or pathological states (e.g., Bence Jones proteins in urine)
- Typical uses include:
- Concentrate proteins, antibody and antigen titers, and enzymes or isoenzymes
- Simple concentration of research samples
- Removal or exchange of salts and other microsolutes by repetitive dilution and concentration
- Purification of membrane-retained macrosolute
- Absorbent pulls solvent and salts through the ultrafilter, concentrating the sample
- Requires no additional equipment and can be operated unattended
- Non-sterile, disposable, 8-well ultrafiltration device with a volume of 5mL (50μL final concentrate volume)
Specifications
Clear | |
Single-use | |
15 kDa | |
22 cm2 | |
40/Pk. | |
5 mL |
14.7cmL x 4.5cmW x 10.2cmH | |
PES | |
Non-sterile | |
Concentration of Urine and Cerebrospinal Fluid for Enrichment of Analytes in Clinical Labs | |
8 Chambers/Unit |