missing translation for 'onlineSavingsMsg'
Learn More

Mineral oil, MP Biomedicals™

£27.79 - £41.77

Chemical Identifiers

CAS 8042-47-5
Molecular Formula C16H10N2Na2O7S2
Molecular Weight (g/mol) 452.363
InChI Key AEOVEGJBKQQFOP-DDVLFWKVSA-L
Synonym acid orange 10, wool orange 2g, orange g, c.i. acid orange 10, c.i. orange g, c.i. food orange 4, light orange g, colacid orange g, dolkwal orange g, hexacol orange g
PubChem CID 9566064
IUPAC Name disodium;(8Z)-7-oxo-8-(phenylhydrazinylidene)naphthalene-1,3-disulfonate
SMILES C1=CC=C(C=C1)NN=C2C(=O)C=CC3=CC(=CC(=C32)S(=O)(=O)[O-])S(=O)(=O)[O-].[Na+].[Na+]
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11452442 View Documents MP Biomedicals
0215169490
500mL £27.79
500mL View alternative products
Save Up to 
»
Estimated Shipment
22-09-2020
Please to purchase this item. Need a web account? Register with us today!
11416562 View Documents MP Biomedicals
0215013880
100mL £41.77
100mL View alternative products
Save Up to 
»
Estimated Shipment
22-09-2020
Please to purchase this item. Need a web account? Register with us today!
11462442 View Documents MP Biomedicals
0215169491
1L £38.70
1 L View alternative products
Save Up to 
»
Estimated Shipment
16-07-2020
Please to purchase this item. Need a web account? Register with us today!
Specifications

Chemical Identifiers

8042-47-5
452.363
acid orange 10, wool orange 2g, orange g, c.i. acid orange 10, c.i. orange g, c.i. food orange 4, light orange g, colacid orange g, dolkwal orange g, hexacol orange g
disodium;(8Z)-7-oxo-8-(phenylhydrazinylidene)naphthalene-1,3-disulfonate
C16H10N2Na2O7S2
AEOVEGJBKQQFOP-DDVLFWKVSA-L
9566064
C1=CC=C(C=C1)NN=C2C(=O)C=CC3=CC(=CC(=C32)S(=O)(=O)[O-])S(=O)(=O)[O-].[Na+].[Na+]

Specifications

Mineral oil
Colorless
8042-47-5
acid orange 10, wool orange 2g, orange g, c.i. acid orange 10, c.i. orange g, c.i. food orange 4, light orange g, colacid orange g, dolkwal orange g, hexacol orange g
C1=CC=C(C=C1)NN=C2C(=O)C=CC3=CC(=CC(=C32)S(=O)(=O)[O-])S(=O)(=O)[O-].[Na+].[Na+]
452.363
Viscous Liquid
0.81 to 0.894 at 15°C
500mL
C16H10N2Na2O7S2
AEOVEGJBKQQFOP-DDVLFWKVSA-L
disodium;(8Z)-7-oxo-8-(phenylhydrazinylidene)naphthalene-1,3-disulfonate
9566064