missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Mimecan Antibody, R&D Systems™
Goat Polyclonal Antibody has been used in 3 publications
POA
Specifications
Antigen | Mimecan/OIF |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 0.1 ug/mL |
Applications | Western Blot |
Classification | Polyclonal |
Description
Mimecan/OIF Polyclonal specifically detects Mimecan/OIF in Mouse samples. It is validated for Western Blot.Specifications
Mimecan/OIF | |
Western Blot 0.1 ug/mL | |
Polyclonal | |
Goat | |
Mouse | |
DKFZp586P2421, KSPG25, mimecan, mimecan proteoglycan, OGN, OIF, OIFOG, osteoglycin, osteoglycin (osteoinductive factor), Osteoinductive factor, SLRR3A, SLRR3Acorneal keratan sulfate proteoglycan | |
OGN | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Western Blot | |
Unconjugated | |
RUO | |
Q62000 | |
4969 | |
Mouse myeloma cell line NS0-derived recombinant mouse Mimecan Ala20-Phe298 Accession # Q62000 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects mouse Mimecan in direct ELISAs and Western blots. In direct ELISAs, approximately 10% cross-reactivity with recombinant human Mimecan is observed. |
For Research Use Only