missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals MIF Antagonist, ISO-1
210.67 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
5 mg
25 mg
Description
MIF Antagonist, ISO-1 inhibits MIF dopachrome tautomerase activity by binding to its catalytic active site (IC50 = 7 μM for D-dopachrome tautomerase) and suppresses the production of TNFα, PGE2 and COX-2 in human monocytes and arachidonic acid in RAW 264.7 macrophages. MIF Antagonist, ISO-1 has been shown to exhibit antidiabetogenic properties in immunoinflammatory diabetic mouse model.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Quantity | 25 mg |
Description | Pale Yellow |
Content And Storage | -30°C to -10°C |
For Use With (Application) | A cell-permeable isoxazoline compound |
Formula Weight | 235.24g/mol |
Sensitivity | Light Sensitive |
Product Type | Signal Transduction Reagent |
Form | Solid |
InChI Key | CLVQGFPQFNASJH-UHFFFAOYNA-N |
SMILES | COC(=O)CC1CC(NO1)=C1C=CC(=O)C=C1 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'