Learn More
MIER3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA566161
Additional Details : Weight : 0.01000kg
Description
Immunogen sequence: HETSDFFPRPL RSNTACDGDK ESEVEDVETD SGNSPEDLRK EIMIGLQYQA EIPPYLGEYD GNEK Highest antigen sequence identity to the following orthologs - mouse 86%, rat 94%.
Transcriptional repressor.Specifications
MIER3 | |
Polyclonal | |
Unconjugated | |
MIER3 | |
5730509D14; AU015422; D130064H19Rik; mesoderm induction early response 1, family member 3; mesoderm induction early response protein 3; MIER family member 3; Mier3; Mi-er3; RGD1311241 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
166968 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q7Z3K6 | |
MIER3 | |
Recombinant Human MIER3 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
For Research Use Only