Learn More
Spectrum Labs™ MidiKros™ TC Filter Module Fittings Kit
Spectrum Labs™ MidiKros™ TC Filter Module Fittings Kit includes a variety of adapters, tubing and other fitting accessories for your filter module.
Brand: Spectrum Labs™ ACPX-T-01N
Additional Details : Weight : 1.00000kg
Description
- 100 White 3.9 in. Cable Ties
- 4 1/5 in. to 3/8 in. OD Tubing Pinch Clamps
- 5 Slide Clamps
- 15 Male Luer Caps, Polypropylene
- 12 Male Luer Lock x 1/16 in. HB Adapters, Polypropylene
- 20 Female Luer Lock x 1/16 in. HB Adapters, Polypropylene
- 12 Male Luer Lock x 3/32 in. HB Adapters, Polypropylene
- 15 Female Luer Lock x 3/32 in. HB Adapters, Polypropylene
- 11 Male Luer Lock x 1/8 in. HB Adapters, Polypropylene
- 14 Female Luer Lock x 1/8 in. HB Adapters, Polypropylene
- 10 Male Luer Lock x 1/4 in. HB Adapters, Polypropylene
- 10 Female Luer Lock x 1/4 in. HB Adapters, Polypropylene
- 8 Female Luer Lock Tees, Polypropylene 1
- 0 Female Luer Lock x Female Luer Lock Couplers
Specifications
Kit for filter module | |
MidiKros TC Filter Module | |
Ties, adapters, gaskets, tubing, couplers, clamps, tabs |